Learn More
Pyrex™ Graduated Dropping Funnel with Glass Key
Used to transfer fluids.
Brand:  Pyrex 3590/04
 View more versions of this product
Product Code. 11313684
  • 148.00 € / Each
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Complies with ISO 4800 type 4
  • Manufactured from Pyrex borosilicate glass
  • Double graduated scale showing approximate volume contained/dispensed
  • Graduations in white enamel
  • With interchangeable glass key
  • Supplied with glass stopper
TRUSTED_SUSTAINABILITY
Specifications
Specifications
250 mL
Yes
5mL
Borosilicate Glass
Glass stopper and key
Dropping Funnel