missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Dessicator Plates

For use with Pyrex vacuum and non-vacuum desiccators

Brand:  Pyrex 1597/04D

62.80 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 11955879

  • 69.20 € / Each
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

For use with Pyrex vacuum and non-vacuum desiccators

  • For use with Pyrex vacuum and non-vacuum desiccators
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Desiccator Plate
Plate Size: 200mm
Metal
Vacuum and non-vacuum desiccators
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers