Learn More
Quickfit™ Borosilicate Glass Digestion Tube with Socket
Designed for use with blockheaters
Brand: Quickfit TJ75/26/3
Code : NEW
Additional Details : Weight : 0.56000kg
Description
- Contains Quickfit precision-ground socket to accept air condensers
- Manufactured from borosilicate glass for highest chemical and thermal durability