Learn More
Corning™ PYREX™ Disposable Individually Wrapped Glass Serological Pipets, Sterile
Calibrated to deliver (TD) with blow-out. Corning™ Pyrex™ Disposable Individually Wrapped Glass Serological Pipets offer long, slender, tapered tips to make pipetting go faster and easier.
Brand:  Corning™ 7077-5N
 View more versions of this product
Product Code. 10494343
  • 380.30 € / Pack of 720
Estimated Shipment: 03-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
These pipets, calibrated 'To Deliver' with blow-out, offer long, slender, tapered tips to make ­pipetting go faster and easier. All pipets have negative graduations and fire-polished tips for ­burr-free, even flow rates. Easy-open Steri-ViewTM wrappers (one side paper, one side plastic) have color-coded sizes printed directly on the paper for easy identification by size. Colors conform to ISO standards. Reference: ASTM E-714.
Do not pipet by mouth. We suggest using a mechanical pipetting device.
Order Info
Individually packaged in easy-open Steri-View™ wrappers or multipacked in plastic bags.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ASTM E714
Yes
1 mL
0.16 oz.
5 mL
Single-use
Glass
0.1 mL
Round
Blue
Serological Pipet