Learn More
Corning™ PYREX™ Disposable Round-Bottom Rimless Glass Tubes
Manufactured from borosilicate glass to reduce pH changes and contaminants. Corning™ PYREX™ Disposable Round-Bottom Rimless Glass Tubes are designed for both tissue culture and general bacteriological work.
Brand:  Corning™ 99445-16XX
 View more versions of this product
Product Code. 10018522
  • 150.00 € / Pack of 1000
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Manufactured from borosilicate glass to reduce pH changes and contaminants. Corning™ PYREX™ Disposable Round-Bottom Rimless Glass Tubes are designed for both tissue culture and general bacteriological work.
These one-use culture tubes are packaged for your convenience in easy-to-open, easy-to-use packs. The three small sizes come in a dispenser arrangement, and the larger sizes are bulk-packed. Made of borosilicate glass to reduce pH changes and contaminants potentially leached from soda-lime glass.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
23 mL
Borosilicate Glass
For tissue culture work and general bacteriological use
Bulk
Non-sterile
16 mm
150 mm
Single-use
None
No
No
Culture Tube
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Information
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ PYREX™ Disposable Round-Bottom Rimless Glass Tubes > 150 mm; 23 mL