missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Funnel Repair Stopcocks with PTFE Key
Contains one flared end for connection to the base of the funnel
72.25 € - 156.00 €
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
13481899
|
Pyrex
SST704 |
Stopcock diameter: 18.8mm; Key bore: 4mm | N/A | ||||||
13491899
|
Pyrex
SST706 |
Stopcock diameter: 18.8mm; Key bore: 6mm | N/A | ||||||
12451021
|
Pyrex
SST703 |
Stopcock diameter: 14.5mm; Key bore: 2.5mm | N/A | ||||||
13471899
|
Pyrex
SST702 |
Stopcock diameter: 12.5mm; Key bore: 1.5mm | N/A | ||||||
Description
- Contains one flared end for connection to the base of the funnel and 100mm long stem with angled tip
- Straight bore
Specifications
Funnel Repair | |
Glass | |
PTFE key |