missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Funnel with Sintered Glass Disc
Manufactured from Pyrex™ Borosilicate Glass. Pyrex™ Funnel with Sintered Glass Disc is used for the filtration of solutions. Resistant to most corrosive agents, and eliminates the need for filter paper.
Brand: Pyrex 3750/03
57.19 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Manufactured from Pyrex™ Borosilicate Glass. Pyrex™ Funnel with Sintered Glass Disc is used for the filtration of solutions. Resistant to most corrosive agents, and eliminates the need for filter paper
- Sintered disc manufactured from Pyrex™ borosilicate glass
- For the filtration of solutions: - No requirement for filter paper - Resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
- Available in a choice of porosities
Specifications
30 mL | |
Borosilicate Glass | |
For filtration of solutions | |
Sintered glass disc | |
3 | |
Filter Funnel |
8mm | |
144 mm | |
30mm | |
82mm | |
1 to 16μm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction