missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Funnel with Sintered Glass Disc
Manufactured from Pyrex™ Borosilicate Glass. Pyrex™ Funnel with Sintered Glass Disc is used for the filtration of solutions. Resistant to most corrosive agents, and eliminates the need for filter paper.
Brand: Pyrex 3730/04
450.30 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Manufactured from Pyrex™ Borosilicate Glass. Pyrex™ Funnel with Sintered Glass Disc is used for the filtration of solutions. Resistant to most corrosive agents, and eliminates the need for filter paper
- Sintered disc manufactured from Pyrex™ borosilicate glass
- For the filtration of solutions: - No requirement for filter paper - Resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
- Available in a choice of porosities
Specifications
20 mL | |
Borosilicate Glass | |
For filtration of solutions | |
Sintered glass disc | |
2 | |
Filter Funnel |
8mm | |
130 mm | |
20mm | |
75mm | |
40 to 100μm |