Learn More
Pyrex™ Gas Distribution Tube
Disperse gas in liquids to produce the optimum bubble size. Pyrex™ gas distribution tubes come in a variety of porosities.
70.70 € - 104.00 €
Specifications
Material Borosilicate Glass (3.3)
Overall Length (Metric) 300 mm
For Use With (Application) For the dispersion of gas in liquids
Includes Sintered thimble
View More Specs
Products 4
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
15608166
View Documents
Pyrex
3830/04M
Porosity: 2
104.00 €
Pack of 2
In Stock
to see stock.
Edge
Add to basket
12663232
View Documents
Pyrex
3830/02M
Porosity: 1
79.85 €
Pack of 2
Estimated Shipment: 19-06-2025
to see stock.
Add to basket
12547866
View Documents
Pyrex
3830/06M
Porosity: 3
70.70 €
Pack of 2
Estimated Shipment: 19-06-2025
to see stock.
Add to basket
10217634
View Documents
Pyrex
3830/00M
Porosity: 0
89.40 €
Pack of 2
Estimated Shipment: 19-06-2025
to see stock.
Add to basket
Description
Description
Disperse gas in liquids to produce the optimum bubble size. Pyrex™ gas distribution tubes come in a variety of porosities.
  • Ideal for the dispersion of gas in liquids
  • Borosilicate glass tube with sintered thimble
  • Choice of thimble porosities to achieve optimum gas bubble size
Specifications
Specifications
Borosilicate Glass (3.3)
300 mm
For the dispersion of gas in liquids
Sintered thimble
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Pyrex™ Gas Distribution Tube