Learn More
Pyrex™ Borosilicate Glass Cylinders in Squat Format
Printed with blue enamel graduations for enhanced visibility. Pyrex™ Borosilicate Glass Cylinders in Squat Form are ideal for use in fume hoods and restricted spaces. Borosilicate glass gives superior chemical resistance.
Brand:  Pyrex 3285-50
77.76 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 12692032
  • 83.90 € / Pack of 2
Estimated Shipment: 15-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Hexagonal base and squat form delivers stability
  • Heavy duty construction increases mechanical strength and reduces breakage (lowering cost and improving user safety)
  • Heavily beaded rim and spout allow clean and easy dispensing
  • Class B
  • Complies with ISO 4788
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Case of 2
Blue
Graduated Cylinder
Borosilicate Glass
B
Confined Spaces
Blue enamel
Hexagonal
Round
±1 mL
BS 604, DIN 12680, ISO 4788
50 mL
148 mm
1 mL