Learn More
PYREX™ Borosilicate Glass Graduated Cylinders
Printed with blue enamel graduations for enhanced visibility. Pyrex™ Borosilicate Glass Cylinders in Tall Form provide superior chemical resistance.
Brand:  Pyrex 3280-500
 View more versions of this product
Product Code. 12672042
  • 182.00 € / Pack of 2
Estimated Shipment: 01-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Printed with blue enamel graduations for enhanced visibility. Pyrex™ Borosilicate Glass Cylinders in Tall Form provide superior chemical resistance.
  • Hexagonal base delivers stability
  • Heavy duty construction increases mechanical strength and reduces breakage (lowering cost and improving user safety)
  • Heavily beaded rim and spout allow clean and easy dispensing
  • Class B
  • Complies with ISO 4788
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hexagonal
Graduated Cylinder
Borosilicate Glass
B
365 mm
5 mL
Blue
±5 mL
BS 604, DIN 12680, ISO 4788
500 mL
Blue enamel