Learn More
Pyrex™ Labware Polyethylene Reservoir Bottle
Feed liquids into a Sofnol burette with easy this filling squeeze bottle. The Pyrex™ labware polyethylene reservoir bottle is a replacement part for the Sofnol, auto zero, picnch clip, class B burette
Brand:  Pyrex 3299/06
24.65 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 11343544
  • 26.60 € / Each
Estimated Shipment: 31-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Polyethylene
  • Squeeze bottle
  • For use with Sofnol burette
  • Squeeze the bottle to fill the burette
  • Release pressure to allow excess liquid to drain back into bottle and set the burrette to zero
  • Used for liquid testing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1000 mL
Sofnol Burette
Polyethylene
Reservoir Bottle