Learn More
Azlon™ Labware Detergent
Contains no chlorine bleaches, phosphates, or enzymes. With excellent cleaning performance, Azlon™ Labware Detergent is ideal for use with laboratory glassware, plasticware, rubber, ceramics and stainless steel. Economically priced, non-corrosive, non-irritant, and totally biodegradable surface-active cleaning agent.
Brand:  Azlon™ 40023
134.40 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Product Code. 12549965
  • 167.00 € / Pack of 2
Edge
Explore more special offers
Description
Description
  • Excellent cleaning performance, ideal for use with laboratory glassware, plasticware, rubber, ceramics and stainless steel
  • Economical price 
  • Non-corrosive, non-irritant, and totally biodegradable surface-active cleaning agent
  • Free of chlorine bleaches, phosphates and enzymes
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Lipsol Detergent
10.78 (at 20°C)
100°C
2 Pk.
5 L
Liquid
Clear
Ideal for use with laboratory Glassware, Plasticware, Rubber, Ceramics and Stainless Steel
169.07oz.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Azlon™ Labware Detergent > 2/Pk.