Learn More
Fisherbrand™ Pyrex™ Magnetic Stir Bar
Useful for high temperature applications up to 274°C. Fisherbrand™ Pyrex™ Magnetic Stir Bars have a completely smooth surface offering zero absorption and are easy to clean.
Brand:  Fisherbrand™ 371015008
Product Code. 15535594
  • 26.15 € / Each
Estimated Shipment: 03-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Autoclavable
  • Glass encapsulated Alnico magnet
  • Manufactured in registered ISO 9001:2008 facilities
  • Meets US Pharmacopoeia specifications for Type I borosilicate glass
Specifications
Specifications
Autoclavable