missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ PYREX™ Media Bottle Septa
Click to view available options
Material:
PTFE-faced Silicone Rubber
Silicone Rubber
For Use With (Equipment):
GL25 open top PBT screw cap
GL32 open top PBT screw cap
GL45 open top PBT screw cap
Description
These are various sizes and types of screw caps for use with storage bottles.
Specifications
Specifications
Color | Natural |
Material | Silicone Rubber |
No. per Case | 10/Cs. |
No. per Pack | 10/Pk. |
Quantity | 10 Cs. |
Sterility | Non-sterile |
For Use With (Equipment) | GL45 open top PBT screw cap |
Closure Size | GL 45 |
Closure Type | No Closure |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'