Learn More
Corning™ PYREX™ Media Bottle Septa
Description
These are various sizes and types of screw caps for use with storage bottles.
Specifications
Specifications
| Color | Natural |
| Material | Silicone Rubber |
| No. per Case | 10/Cs. |
| No. per Pack | 10/Pk. |
| Quantity | 10 Cs. |
| Sterility | Non-sterile |
| For Use With (Equipment) | GL45 open top PBT screw cap |
| Closure Size | GL 45 |
| Closure Type | No Closure |
Frequently Asked Questions (FAQs)
Proper care includes:
• Avoiding mechanical damage (e.g., scratches or chips)
• Preventing thermal shock
• Following recommended cleaning and sterilization techniques
1. Wash bottles promptly after use, or soak them in water if immediate cleaning isn’t possible
2. Sterilize bottles contaminated with biological materials before cleaning
3. Use detergent and rinse thoroughly with tap water, followed by deionized or distilled water
4. For new bottles used in cell culture, soak in a 1% hydrochloric or nitric acid solution before washing
• Do not tighten caps immediately after autoclaving to prevent vacuum breakage, except for vented caps (e.g., 1395-45LTMC)
• Replace vented caps after 3-5 autoclave cycles or if the membrane is damaged
Do not use bottle-top filter units with reagent bottles larger than 1000 mL or laboratory bottles larger than 2000 mL.
PYREX bottles larger than 2000 mL are not recommended for vacuum applications. Additionally, 1396 and 1397 series bottles are not designed for vacuum use.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.