Learn More
PYREX™ Pear-Shaped Glass Separating Funnel with PTFE Key
Manufactured from Pyrex™ glass for corrosion-free performance. Pyrex™ Pear-Shaped Glass Separating Funnel with Glass Key complies with ISO 4800 Type 2 Standards. Includes glass stopper and interchangeable PTFE key.
Brand: Pyrex 2180/06M
112.10 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Description
Manufactured from Pyrex™ glass for corrosion-free performance. Pyrex™ Pear-Shaped Glass Separating Funnel with Glass Key complies with ISO 4800 Type 2 Standards. Includes glass stopper and interchangeable PTFE key.
- Complies with ISO 4800 Type 2
- Manufactured from Pyrex™ glass for corrosion-free performance
- With interchangeable PTFE key
- Supplied with glass stopper

Specifications
250 mL | |
ISO 4800 | |
2.5mm; Glass | |
24/29 |
Borosilicate Glass | |
1PK | |
Glass | |
Separatory Funnel |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.