missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Plastic Coated Borosilicate Glass Wide Neck Round Media Bottles Without Cap

Product Code. 11962629
Click to view available options
Capacity (Metric):
10,000 mL
100 mL
1000 mL
2000 mL
250 mL
500 mL
5000 mL
Unit Size:
Each
Pack of 10
This item is not returnable. View return policy

Product Code. 11962629

Brand: Pyrex 1518/06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Manufactured from Borosilicate glass with clear plastic coating to provide added protection against mechanical breakage. Pyrex™ Plastic Coated Borosilicate Glass Wide Neck Round Media Bottles Without Cap reduces the hazard of flying glass and liquid splashes in the event of breakage. Coating withstands steam sterilisation up to 135° C. Bottles are supplied without cap or pouring ring.

  • Bottles only. Supplied without cap or pouring ring
  • Borosilicate glass with clear plastic coating to provide added protection against mechanical breakage
  • Reduces the hazard of flying glass and liquid splashes in the event of breakage
  • Printed with retrace code: downloadable batch certificate
  • Coating withstands steam sterilisation up to 135°C
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 250 mL
Coating Plastic
Color Clear
For Use With (Application) Ideal for mixing and storage of culture medium
Material Borosilicate Glass
Certifications/Compliance ISO 4796
Graduated Yes
Mouth Wide
Shape Round
Height (Metric) 138 mm
Closure Size 45 mm
Diameter (Metric) 70 mm
Autoclavable Autoclavable
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.