Learn More
Pyrex™ Separating Funnel with Long Delivery Stem and PTFE Key
Supplied with hollow glass stopper and interchangeable PTFE key for grease-free performance. Pyrex™ Separating Funnel with Long Delivery Stem and PTFE Key provides good control of the liquid/liquid interface.
98.95 € - 305.00 €
Specifications
Material Borosilicate Glass
Includes Hollow Glass stopper, PTFE key
Product Type Separatory Funnel
View More Specs
Products 6
Product Code Brand Capacity (Metric) Joint Number Stopcock Price Quantity & Availability  
Product Code Brand Capacity (Metric) Joint Number Stopcock Price Quantity & Availability  
12539165
View Documents
Pyrex
443047N
250 mL 24/29 (Bore Size) 2.5mm
133.00 €
Each
Estimated Shipment: 26-03-2025
to see stock.
Add to basket
12519165
View Documents
Pyrex
443042N
125 mL 19/26 (Bore Size) 1.5mm
110.00 €
Each
Estimated Shipment: 26-03-2025
to see stock.
Add to basket
12559165
View Documents
Pyrex
443054N
500 mL 24/29 (Bore Size) 4mm
162.00 €
Each
Estimated Shipment: 26-03-2025
to see stock.
Add to basket
12599165
View Documents
Pyrex
443065N
2000 mL 29/32 (Bore Size) 4mm
305.00 €
Each
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
12579165
View Documents
Pyrex
443060N
1000 mL 29/32 (Bore Size) 4mm
203.00 €
Each
Estimated Shipment: 26-03-2025
to see stock.
Add to basket
12505067
View Documents
Pyrex
443038N
60 mL 14/23 (Bore Size) 1.5mm
98.95 €
Each
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
Description
Description
  • Gilson style with long delivery stem for good control of the liquid/liquid interface
  • With interchangeable PTFE key for grease-free performance
  • Supplied with hollow glass stopper
Specifications
Specifications
Borosilicate Glass
Hollow Glass stopper, PTFE key
Separatory Funnel