missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Short Stem Glass Funnel
Manufactured from chemical and heat resistant Pyrex™ Borosilicate Glass. Pyrex™ Short Stem Glass Funnel with 60° bowl angle provides precise fitting of filter paper.
Brand: Pyrex 2140/12M
This item is not returnable.
View return policy
Description
Manufactured from chemical and heat resistant Pyrex™ Borosilicate Glass. Pyrex™ Short Stem Glass Funnel with 60° bowl angle provides precise fitting of filter paper.
- With 60° bowl angle for precise fitting of filter paper
- Manufactured from chemical and heat resistant Pyrex™ borosilicate glass
Specifications
150mm | |
Borosilicate Glass | |
NF B 35 032 | |
154mm |
16mm | |
270 mm | |
240 to 270mm (Filter paper size) | |
Short stem funnel |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur