missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-3-carboxamidine hydrochloride 0.5 hydrate, 95%, Thermo Scientific™

Product Code. 10118164
Click to view available options
Quantity:
5 g
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 871825-82-0
Molecular Formula C12H18Cl2N6O
Molecular Weight (g/mol) 333.217
InChI Key AOHLMHDFGXVZAQ-UHFFFAOYSA-N
Synonym pyridine-3-carboximidamide hemihydrate hydrochloride
PubChem CID 90477204
IUPAC Name pyridine-3-carboximidamide;hydrate;dihydrochloride
SMILES C1=CC(=CN=C1)C(=N)N.C1=CC(=CN=C1)C(=N)N.O.Cl.Cl

Specifications

Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 166.61
Percent Purity 95%
Chemical Name or Material Pyridine-3-carboxamidine hydrochloride 0.5 hydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.