missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-4-boronic acid, 97%, Thermo Scientific™

Product Code. 10337853
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 1692-15-5
Molecular Formula C5H6BNO2
Molecular Weight (g/mol) 122.92
MDL Number MFCD01074545
InChI Key QLULGIRFKAWHOJ-UHFFFAOYSA-N
Synonym pyridine-4-boronic acid, 4-pyridineboronic acid, 4-pyridylboronic acid, pyridin-4-yl boronic acid, 4-pyridinylboronic acid, pyridin-4-yl-4-boronic acid, 4-pyridinebornic acid, pyridyl-4-boronic acid, 4-pyridinyl boronic acid, pyridine-4-ylboronic acid
PubChem CID 2734379
SMILES OB(O)C1=CC=NC=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name (pyridin-4-yl)boronic acid
Formula Weight 122.92
Percent Purity 97%
Chemical Name or Material Pyridin-4-ylboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'