missing translation for 'onlineSavingsMsg'
Learn More
Pyridine-4-boronic acid hydrochloride, 95%, Thermo Scientific Chemicals Product Code.: 15489397

Pyridine-4-boronic acid hydrochloride, 95%, Thermo Scientific Chemicals

Product Code. 15489397
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15489397

Brand: Thermo Scientific Alfa Aesar H52648.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 913835-65-1
Molecular Formula C5H7BClNO2
Molecular Weight (g/mol) 159.38
MDL Number MFCD06201888
InChI Key NCZXYQJUECNKAD-UHFFFAOYSA-N
Synonym pyridine-4-boronic acid hydrochloride, pyridin-4-ylboronic acid hydrochloride, pyridine-4-boronic acid, hcl, 4-pyridineboronic acid, hydrochloride, pyridine-4-boronic acid hcl, pyridine-4-boronic acid-hydrogen chloride, boronicacid, b-4-pyridinyl-, hydrochloride 1:1, c5h6bno2.hcl, pyridineboronic acid, 4-, hcl, 4-pyridineboronic acid, hcl
PubChem CID 44118740
IUPAC Name pyridin-4-ylboronic acid;hydrochloride
SMILES Cl.OB(O)C1=CC=NC=C1

Specifications

Melting Point 340°C
Quantity 5 g
Formula Weight 159.38
Percent Purity 95%
Chemical Name or Material Pyridine-4-boronic acid hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pyridine-4-boronic acid hydrochloride, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.