missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridine-4-carboximidamide hydrochloride, 95%, Thermo Scientific™
Click to view available options
Quantity:
5 g
10 g
50 g
Specifications
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Quantity | 5 g |
Formula Weight | 157.6 |
Percent Purity | 95% |
Chemical Name or Material | Pyridine-4-carboximidamide hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction