missing translation for 'onlineSavingsMsg'
Learn More

Pyridine, 99+%, for analysis, Thermo Scientific Chemicals

Product Code. 10341054
Click to view available options
Quantity:
100 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-86-1
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.1
MDL Number MFCD00011732
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
Synonym azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
PubChem CID 1049
ChEBI CHEBI:16227
IUPAC Name pyridine
SMILES C1=CC=NC=C1

Specifications

Melting Point -42.0°C
Density 0.9780g/mL
Boiling Point 115.0°C to 116.0°C
Flash Point 17°C
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.5085 to 1.5105
Quantity 100 mL
Beilstein 20,181
Fieser 01,958; 02,349; 04,414; 06,497; 11,448; 12,416; 13,293; 15,1; 17,153
Merck Index 15,8081
Specific Gravity 0.978
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and most other common, organic solvents
Viscosity 0.95 mPa.s (20°C)
Formula Weight 79.1
Percent Purity 99+%
Grade Analysis
Physical Form Liquid
Chemical Name or Material Pyridine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'