Learn More
Pyridine, 99+%, extra pure, Thermo Scientific Chemicals
22.31 € - 758.78 €
Chemical Identifiers
CAS | 110-86-1 |
---|---|
Molecular Formula | C5H5N |
Molecular Weight (g/mol) | 79.102 |
MDL Number | MFCD00011732 |
InChI Key | JUJWROOIHBZHMG-UHFFFAOYSA-N |
Synonym | azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717 |
PubChem CID | 1049 |
ChEBI | CHEBI:16227 |
IUPAC Name | pyridine |
SMILES | C1=CC=NC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10644802
|
Thermo Scientific Acros
131780500 |
50 mL | Glass Bottle |
22.31 €
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10081420
|
Thermo Scientific Acros
131780010 |
1 L | EcoSafPak™ Glass Bottle |
107.54 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10634802
|
Thermo Scientific Acros
131780025 |
2.5 L | Glass Bottle |
248.24 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10092583
|
Thermo Scientific Acros
131780250 |
25 L | Metal Drum |
758.78 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
110-86-1 | |
79.102 | |
JUJWROOIHBZHMG-UHFFFAOYSA-N | |
1049 | |
pyridine |
C5H5N | |
MFCD00011732 | |
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717 | |
CHEBI:16227 | |
C1=CC=NC=C1 |
Specifications
-42.0°C | |
0.9780g/mL | |
17°C | |
Liquid | |
99+% | |
0.978 | |
MFCD00011732 | |
0.95 mPa.s (20°C) | |
0.1% max. (Coulometric) | |
15, 8081 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and most other common, organic solvents | |
C1=CC=NC=C1 | |
79.102 | |
CHEBI:16227 | |
≥99% | |
Pyridine |
110-86-1 | |
115.0°C to 116.0°C | |
Authentic | |
1.5085 to 1.5105 | |
0.02% max. | |
C5H5N | |
Vapor Pressure: 20mbar at 20°C | |
20, 181 | |
01,958; 02,349; 04,414; 06,497; 11,448; 12,416; 13,293; 15,1; 17,153 | |
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717 | |
JUJWROOIHBZHMG-UHFFFAOYSA-N | |
pyridine | |
1049 | |
79.1 | |
Extra Pure |
Safety and Handling
GHS P Statement
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
- Pyridine
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
RUO – Research Use Only