missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridine, Certified AR for Analysis, Fisher Chemical™
C5H5N, CAS Number-110-86-1, azabenzene, azine, caswell no. 717, piridina, piridina italian, pirydyna, pirydyna polish, pyridin, pyridin german, rcra waste number u196, 2.5L, 115 deg.C, CHEBI:16227, Colorless, 79.1g/mol, JUJWROOIHBZHMG-UHFFFAOYSA-N, pyridine, -42 deg.C, 11732, 79.102
110.00 € - 1620.00 €
Chemical Identifiers
CAS | 110-86-1 |
---|---|
Molecular Formula | C5H5N |
Molecular Weight (g/mol) | 79.102 |
MDL Number | 11732 |
InChI Key | JUJWROOIHBZHMG-UHFFFAOYSA-N |
Synonym | azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717 |
PubChem CID | 1049 |
ChEBI | CHEBI:16227 |
IUPAC Name | pyridine |
SMILES | C1=CC=NC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10102110
|
Fisher Chemical
P/7960/08 |
500 mL | Amber Glass Bottle |
137.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10124433
|
Fisher Chemical
P/7960/PB08 |
500 mL | Plastic-coated Glass Bottle |
110.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10650813
|
Fisher Chemical
P/7960/15 |
1 L | Amber Glass Bottle |
198.00 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10112060
|
Fisher Chemical
P/7960/PB15 |
1 L | Plastic-coated Glass Bottle |
174.00 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10326340
|
Fisher Chemical
P/7960/PB17 |
2.5 L | Plastic-coated Glass Bottle |
1220.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10419570
|
Fisher Chemical
P/7960/25 |
25 L | Metal Can |
1620.00 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
C5H5N, CAS Number-110-86-1, azabenzene, azine, caswell no. 717, piridina, piridina italian, pirydyna, pirydyna polish, pyridin, pyridin german, rcra waste number u196, 2.5L, 115 deg.C, CHEBI:16227, Colorless, 79.1g/mol, JUJWROOIHBZHMG-UHFFFAOYSA-N, pyridine, -42 deg.C, 11732, 79.102
Chemical Identifiers
110-86-1 | |
79.102 | |
JUJWROOIHBZHMG-UHFFFAOYSA-N | |
1049 | |
pyridine |
C5H5N | |
11732 | |
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717 | |
CHEBI:16227 | |
C1=CC=NC=C1 |
Specifications
8.5 | |
110-86-1 | |
115°C | |
C5H5N | |
20mbar at 20°C | |
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717 | |
C1=CC=NC=C1 | |
79.102 | |
CHEBI:16227 |
-42°C | |
Colorless | |
Liquid | |
11732 | |
0.95 mPaS at 20°C | |
JUJWROOIHBZHMG-UHFFFAOYSA-N | |
pyridine | |
1049 | |
79.1g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Pyridine
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title