missing translation for 'onlineSavingsMsg'
Learn More
Pyridine, CHROMASOLV™ Plus, for HPLC, ≥99.9%, Honeywell Riedel-de Haën™ Product Code.: 15642840

Pyridine, CHROMASOLV™ Plus, for HPLC, ≥99.9%, Honeywell Riedel-de Haën™

Product Code. 15642840
1 L, Glass Bottle
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 15642840

Brand: Honeywell-Chromasolv 2704071L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CHROMASOLV™ Plus, for HPLC,≥99.9%

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-86-1
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.102
MDL Number MFCD00011732
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
Synonym azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717Show MoreShow Less
PubChem CID 1049
ChEBI CHEBI:16227
IUPAC Name pyridine
SMILES C1=CC=NC=C1

Specifications

Melting Point -42°C (literature)
Absorbance max. 0.01 at 400 nm, max. 0.01 at 350 nm, max. 0.02 at 335 nm, max. 0.15 at 315 nm, max. 1.0 at 305 nm
CAS Max % 100.0000%
Density 0.978 g/mL at 25 °C(lit.)
Boiling Point 115°C
Quantity 1 L
Flash Point 20°C
Packaging Glass Bottle
Linear Formula C5H5N
UN Number UN1282
Beilstein 103233
Formula Weight 79.1
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material Pyridine, CHROMASOLV™ Plus, for HPLC, ≥99.9%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Pyridine
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302 + H312 + H332-Harmful if swallowed, in contact with skin or if inhaled.
Precautionary Statement
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pyridine, CHROMASOLV™ Plus, for HPLC, ≥99.9%, Honeywell Riedel-de Haën™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.