missing translation for 'onlineSavingsMsg'
Learn More

Pyridine, Extra Pure, SLR, Fisher Chemical™

73.50 € - 1060.00 €

Chemical Identifiers

CAS 110-86-1
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.102
MDL Number 11732
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
Synonym azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
PubChem CID 1049
ChEBI CHEBI:16227
IUPAC Name pyridine
SMILES C1=CC=NC=C1
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10589570
View Documents Promotion Details
Fisher Chemical
P/7920/08
500 mL Amber Glass Bottle
73.50 €
500mL
Please to purchase this item. Need a web account? Register with us today!
10171960
View Documents Promotion Details
Fisher Chemical
P/7920/PB17
2.5 L Plastic-coated Glass Bottle
187.00 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10274630
View Documents Promotion Details
Fisher Chemical
P/7920/21
5 L Metal Can
556.00 €
5 L
Please to purchase this item. Need a web account? Register with us today!
10677032
View Documents Promotion Details
Fisher Chemical
P/7920/25
25 L Metal Can
1060.00 €
25 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

110-86-1
79.102
JUJWROOIHBZHMG-UHFFFAOYSA-N
1049
pyridine
C5H5N
11732
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
CHEBI:16227
C1=CC=NC=C1

Specifications

8.5
110-86-1
115°C
C5H5N
20mbar at 20°C
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
C1=CC=NC=C1
79.102
CHEBI:16227
-42°C
Colorless
Liquid
11732
0.95 mPaS at 20°C
JUJWROOIHBZHMG-UHFFFAOYSA-N
pyridine
1049
79.1g/mol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Pyridine

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications