missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridine hydrobromide, 98%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
Unit Size:
250g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tekniske data
Tekniske data
| CAS | 18820-82-1 |
| Melting Point | 217°C to 220°C |
| Assay Percent Range | 97.5% min. (Argentometry) |
| Molecular Formula | C5H5N·BrH |
| Quantity | 250 g |
| Synonym | pyridine hydrobromide, pyridinium bromide, pyridine, hydrobromide, pyridine, hydrobromide 1:1, pyridine hbr, pyridinehydrobromide, acmc-1bovh, ksc178o2t, pyridine hydrobromide, purum at |
| InChI Key | BBFCIBZLAVOLCF-UHFFFAOYSA-N |
| SMILES | C1=CC=NC=C1.Br |
| IUPAC Name | pyridine;hydrobromide |
| Molecular Weight (g/mol) | 160.01 |
| Vis mere |
Sikkerhed og håndtering
Produktidentifikator
- Pyridine hydrobromide
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion