missing translation for 'onlineSavingsMsg'
Learn More

Pyridine, puriss. p.a., ACS reagent, Honeywell

Product Code. 15634010
Click to view available options
Quantity:
500 mL
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 110-86-1
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.102
MDL Number MFCD00011732
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
Synonym azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
PubChem CID 1049
ChEBI CHEBI:16227
IUPAC Name pyridine
SMILES C1=CC=NC=C1

Specifications

Density 0.978g/mL at 25°C
Quantity 500 mL
Explosion Limit 0.124
Flash Point 17°C (62.6°F)
Ignition Point 899°F
Vapor Pressure 20 mmHg (25°C), 10 mmHg (13.2°C)
UN Number UN1282
Beilstein 103233
Formula Weight 79.1g/mol
Percent Purity ≥99.5% (GC)
Grade ACS/puriss. p.a.
Chemical Name or Material Pyridine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'type03FormHeader'

missing translation for 'productTitle'

missing translation for 'thankYou' missing translation for 'feedbackSubmitted'