missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridinium dichromate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent

Specifications
Specifications
Melting Point | 152.0°C to 153.0°C |
Color | Orange to Brown |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (Iodometry) |
Packaging | Plastic bottle |
Quantity | 50 g |
Fieser | 09,399; 10,335; 12,419; 14,269; 15,57 |
Merck Index | 15,8087 |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in dmf, dmso and dimethylacetamide -fairly, soluble in acetonenitrile - sparingly soluble in, methylene chloride, chloroform and ether - pract., insol.in hexane,toluene,ethyl ether,ethyl acetate |
Formula Weight | 376.2 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0437156 | Certificate of Analysis | 25/08/2023 | 200882500,200880500,200880010 |
A0402789 | Certificate of Analysis | 14/12/2022 | 200882500,200880500,200880010 |
A0395750 | Certificate of Analysis | 14/12/2022 | 200882500,200880500,200880010 |
A0408434 | Certificate of Analysis | 13/12/2022 | 200882500,200880500,200880010 |
A0447156 | Certificate of Analysis | 29/11/2022 | 200882500,200880500,200880010 |
Safety and Handling





Product Identifier
- Pyridinium dichromate
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Flammable solid Category 1
- Oxidising solid Category 2
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H314-Causes severe skin burns and eye damage.
- H350i-May cause cancer by inhalation.
- H272-May intensify fire; oxidiser.
- H228-Flammable solid.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Pyridinium dichromate, 98%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction