missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrido[3,2-d][1,3]thiazole-2-thiol, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 168.24 |
Percent Purity | 95% |
Chemical Name or Material | Pyrido[3, 2-d][1, 3]thiazole-2-thiol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'