missing translation for 'onlineSavingsMsg'
Learn More

Pyrido[3,2-d][1,3]thiazole-2-thiol, 97%, Thermo Scientific™

Product Code. 10531322
1 g, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 57135-09-8
Molecular Formula C6H4N2S2
Molecular Weight (g/mol) 168.232
MDL Number MFCD00178760
InChI Key WITYIAHCBUYCSY-UHFFFAOYSA-N
Synonym thiazolo 5,4-b pyridine-2 1h-thione, thiazolo 5,4-b pyridine-2-thiol, 1,3 thiazolo 5,4-b pyridine-2-thiol, pyrido 3,2-d 1,3 thiazole-2-thiol, 2-mercaptothiazolo 5,4-b pyridine, pyrido 3,2-d-1,3-thiazol-2-thiol, 2-mercapto-1,3 thiazolo 5,4-b pyridine, 2-sulphanyl-1,3 thiazolo 5,4-b pyridine, 1h-1,3 thiazolo 5,4-b pyridine-2-thione, thiazolo 5,4-b pyridine-2-thione
PubChem CID 2782239
IUPAC Name 1H-[1,3]thiazolo[5,4-b]pyridine-2-thione
SMILES C1=CC2=C(N=C1)SC(=S)N2

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 168.24
Percent Purity 95%
Chemical Name or Material Pyrido[3, 2-d][1, 3]thiazole-2-thiol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pyrido[3,2-d][1,3]thiazole-2-thiol, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.