missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Pyridoxal 5-phosphate monohydrate, 99% Product Code.: 10249693

Thermo Scientific Chemicals Pyridoxal 5-phosphate monohydrate, 99%

Product Code. 10249693
25 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10249693

Brand: Thermo Scientific Chemicals 228170250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41468-25-1
Molecular Formula C8H10NO6P
Molecular Weight (g/mol) 247.14
MDL Number MFCD00006333
InChI Key NGVDGCNFYWLIFO-UHFFFAOYSA-N
Synonym 4-formyl-5-hydroxy-6-methylpyridin-3-yl methyl dihydrogen phosphate hydrate, pyridoxal phosphate hydrate, pyridoxal 5-phosphate monohydrate, phosphopyridoxal monohydrate, pyridoxyl phosphate monohydrate, unii-5v5ioj8338, pyridoxal-5-phosphate, monohydrate, pyridoxal-5'-phosphate monohydrate, pyridoxal-5-phosphate monohydrate, pyridoxal 5'-phosphate monohydrate
PubChem CID 38882
SMILES CC1=NC=C(COP(O)(O)=O)C(C=O)=C1O

Specifications

Melting Point 140.0°C to 143.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Merck Index 10, 7879
Solubility Information Solubility in water: 5g/L water (20°C).
IUPAC Name [(4-formyl-5-hydroxy-6-methylpyridin-3-yl)methoxy]phosphonic acid hydrate
Formula Weight 265.16
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Pyridoxal 5-phosphate monohydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0379464 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number228170050,228170250,228170010
Lot NumberA0453014 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number228170050,228170250,228170010
Lot NumberA0451067 Certificate TypeCertificate of Analysis Date19/04/2023 Catalog Number
Lot NumberA0400893 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number228170050,228170250,228170010
Lot NumberA0409457 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number228170050,228170250,228170010

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Pyridoxal 5-phosphate monohydrate, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.