Learn More
Lonza™ Pyrogen-free Test Tubes
Depyrogenated at extremely high temperatures well in excess of standard depyrogenation process conditions.
Brand:  Lonza™ N205
Product Code. 11610311
  • 137.00 € / Each
Estimated Shipment: 21-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Pyrogen-free borosilicate glass test tubes without screw-caps.
  • Recommended for gel-clot testing.
  • Three packages with single vials front view.
  • Suitable for use when performing the bacterial endotoxins test (BET).
  • Certified to contain less than 0.005 EU/mL endotoxin.
  • All test tubes are made from USP Type 1 flint borosilicate glass.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Type 1 flint borosilicate glass
Borosilicate Glass
10 mm
75 mm
50 Tubes
Test Tube