Learn More
Lonza™ PYROGENT™-5000 Kinetic Turbidimetric LAL Assay Kits
Detection of Gram-negative bacterial endotoxin. Lonza PYROGENT™-5000 Kinetic Turbidimetric LAL Assay Kits is ideal for water samples, large volume parenterals and water rinse from medical devices.
Brand:  Lonza™ N688
Product Code. 11640331
  • 22590.00 € / Pack of 4500
Explore more special offers
Description
Description
  • Ideal for large numbersof samples
  • Sample mixed with reconstituted LAL reagent in 96-well plate
  • Placed in incubating absorbance plate reader
  • Automatically monitored over time for appearance of turbidity
  • Presence of endotoxin lysate will begin to gel causing solution to become cloudy or turbid
TRUSTED_SUSTAINABILITY
Specifications
Specifications
4500 Tests
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lonza™ PYROGENT™-5000 Kinetic Turbidimetric LAL Assay Kits > 4500 tests