missing translation for 'onlineSavingsMsg'
Learn More

Pyrophosphoryl chloride, Thermo Scientific Chemicals

Product Code. 11348027
Click to view available options
Quantity:
0.5 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Pyrophosphoryl chloride serves as a reductive chlorination reagent used in the synthesis of quetiapine. It acts as a phosphorylating agent involved in nucleoside synthesis. It plays an important role in Vilsmeier formylation and glyoxylation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13498-14-1
Molecular Formula Cl4O3P2
Molecular Weight (g/mol) 251.745
MDL Number MFCD00061542
InChI Key CNTIXUGILVWVHR-UHFFFAOYSA-N
Synonym diphosphoryl chloride, pyrophosphoryl chloride, diphosphoryl tetrachloride, diphosphoric tetrachloride, pyrophosphoric tetrachloride, dichlorophosphoryl oxyphosphonoyl dichloride, pyrophosphoryl tetrachloride, pyrophosphorylchloride, phosphorodichloridic anhydride
PubChem CID 4432411
SMILES O=P(OP(=O)(Cl)Cl)(Cl)Cl

Specifications

Boiling Point 101°C (10mmHg)
Melting Point -17°C
Color Colorless through Yellow
Physical Form Liquid
Quantity 5 g
UN Number UN3264
Solubility Information Reacts violently with water
Formula Weight 251.76
Odor Acrid
Sensitivity Air & Moisture Sensitive
Density 1.82g/mL
Chemical Name or Material Pyrophosphoryl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'