missing translation for 'onlineSavingsMsg'
Learn More

Pyrrole-2-carboxaldehyde, 99%, Thermo Scientific Chemicals

Product Code. 11453990
Click to view available options
Quantity:
10 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Pyrrole-2-carboxaldehyde, 99%, C5H5NO, CAS Number-1003-29-8, 2-pyrrolecarboxaldehyde, 2-pyrrolecarbaldehyde, 2-pyrrolaldehyde, 1h-pyrrole-2-carboxaldehyde, alpha-pyrrolaldehyde, 2-formylpyrrole, 2-pyrrolylcarboxaldehyde, pyrrole-2-aldehyde, pyrrole-2-carboxaldehyde, pyrrole-2-carbaldehyde, 100g

Pyrrole-2-carboxaldehyde is used as a pharmaceutical intermediate. It undergoes condensation reaction to produce pyrrole-2-carboxaldehyde salicylhydrazone (PCSH). It is used as a key starting material for the preparation of pyrrole -2-carboxylic acid by oxidation reaction. It is also used to prepare 4-Difluoro-4-bora-3a,4a-diaza-s-indacene (BODIPY dyes) through condensation with a pyrrole, which is an important probes for biotechnology and other applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrrole-2-carboxaldehyde is used as a pharmaceutical intermediate. It undergoes condensation reaction to produce pyrrole-2-carboxaldehyde salicylhydrazone (PCSH). It is used as a key starting material for the preparation of pyrrole -2-carboxylic acid by oxidation reaction. It is also used to prepare 4-Difluoro-4-bora-3a,4a-diaza-s-indacene (BODIPY dyes) through condensation with a pyrrole, which is an important probes for biotechnology and other applications.

Solubility
Soluble in chloroform, dimethyl sulfoxide and methanol. Insoluble in water.

Notes
Store in cool place. Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong reducing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1003-29-8
Molecular Formula C5H5NO
Molecular Weight (g/mol) 95.101
MDL Number MFCD00005217
InChI Key ZSKGQVFRTSEPJT-UHFFFAOYSA-N
Synonym pyrrole-2-carboxaldehyde, 2-formylpyrrole, 1h-pyrrole-2-carboxaldehyde, pyrrole-2-aldehyde, 2-pyrrolecarboxaldehyde, pyrrole-2-carbaldehyde, 2-pyrrolecarbaldehyde, 2-pyrrolylcarboxaldehyde, 2-pyrrolaldehyde, alpha-pyrrolaldehyde
PubChem CID 13854
ChEBI CHEBI:59978
IUPAC Name 1H-pyrrole-2-carbaldehyde
SMILES C1=CNC(=C1)C=O

Specifications

Melting Point 40°C to 46°C
Boiling Point 217°C to 219°C
Flash Point 106°C (222°F)
Quantity 25 g
Beilstein 105745
Sensitivity Air sensitive
Solubility Information Soluble in chloroform,dimethyl sulfoxide and methanol. Insoluble in water.
Formula Weight 95.1
Percent Purity 99%
Chemical Name or Material Pyrrole-2-carboxaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'