missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrrole-3-carboxylic acid hydrate, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 144.0°C to 147.0°C |
Color | Beige to Brown |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min. (HPLC) |
Packaging | Glass bottle |
Quantity | 2.5 g |
Beilstein | 22,II,17 |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in benzene, diethyl ether, petrol. ether |
Formula Weight | 111.1 |
Percent Purity | 95% |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 9
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Pyrrole-3-carboxylic acid hydrate, 95%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction