missing translation for 'onlineSavingsMsg'
Learn More

Pyrrole, 99%, extra pure, Thermo Scientific Chemicals

Product Code. 10011840
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-97-7
Molecular Formula C4H5N
Molecular Weight (g/mol) 67.09
MDL Number MFCD00005216
InChI Key KAESVJOAVNADME-UHFFFAOYSA-N
Synonym pyrrole, divinylenimine, azole, imidole, pyrrol, monopyrrole, divinyleneimine, 1-aza-2,4-cyclopentadiene, polypyrrole, unii-86s1zd6l2c
PubChem CID 8027
ChEBI CHEBI:19203
IUPAC Name 1H-pyrrole
SMILES N1C=CC=C1

Specifications

Melting Point -23.0°C
Color Brown to Colorless
Density 0.9600g/mL
Boiling Point 129.0°C to 131.0°C
Flash Point 36°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5080 to 1.5100
Quantity 25 g
Beilstein 20,159
Merck Index 15,8128
Specific Gravity 0.96
Solubility Information Solubility in water: 60g/L (20°C). Other solubilities: insoluble in dilute alkalies, soluble in alcohol, ether and dilute acids, soluble in most organic solvents
Viscosity 1.317 mPa.s (20°C)
Formula Weight 67.09
Percent Purity 99%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Pyrrole
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Pyrrole
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'