missing translation for 'onlineSavingsMsg'
Learn More

Pyrrolidine, 99%, Thermo Scientific Chemicals

28.64 € - 231.15 €

Chemical Identifiers

CAS 123-75-1
Molecular Formula C4H10ClN
Molecular Weight (g/mol) 107.58
MDL Number MFCD00005249
InChI Key FCLZCOCSZQNREK-UHFFFAOYSA-N
Synonym tetrahydropyrrole, tetrahydro pyrrole, azacyclopentane, azolidine, tetramethylenimine, butylenimine, perhydropyrrole, prolamine, 1-azacyclopentane, tetramethyleneimine
PubChem CID 31268
ChEBI CHEBI:33135
IUPAC Name hydrogen pyrrolidine chloride
SMILES [H+].[Cl-].C1CCNC1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11412637
View Documents
Thermo Scientific Alfa Aesar
A14852.AE
100 mL
28.64 €
100mL
Please to purchase this item. Need a web account? Register with us today!
11422637
View Documents
Thermo Scientific Alfa Aesar
A14852.AP
500 mL
54.67 €
500mL
Please to purchase this item. Need a web account? Register with us today!
11432637
View Documents
Thermo Scientific Alfa Aesar
A14852.0F
2500 mL
231.15 €
2500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Pyrrolidine is used as building block in the synthesis of organic compounds. It is used to activate ketones and aldehydes toward nucleophilic addition. It is also utilized in promoting aldol condensation of ketones and aldehydes by forming enamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrrolidine is used as building block in the synthesis of organic compounds. It is used to activate ketones and aldehydes toward nucleophilic addition. It is also utilized in promoting aldol condensation of ketones and aldehydes by forming enamine.

Solubility
Miscible with alcohol, ether, chloroform and water.

Notes
Store in cool place. Incompatible with acid chlorides, acid anhydrides, strong oxidizing agents, carbon dioxide and acids.
Specifications

Chemical Identifiers

123-75-1
107.58
FCLZCOCSZQNREK-UHFFFAOYSA-N
31268
hydrogen pyrrolidine chloride
C4H10ClN
MFCD00005249
tetrahydropyrrole, tetrahydro pyrrole, azacyclopentane, azolidine, tetramethylenimine, butylenimine, perhydropyrrole, prolamine, 1-azacyclopentane, tetramethyleneimine
CHEBI:33135
[H+].[Cl-].C1CCNC1

Specifications

123-75-1
0.856
2°C (35°F)
Like Rotten Eggs
1.4431
UN1922
Air Sensitive
tetrahydropyrrole, tetrahydro pyrrole, azacyclopentane, azolidine, tetramethylenimine, butylenimine, perhydropyrrole, prolamine, 1-azacyclopentane, tetramethyleneimine
FCLZCOCSZQNREK-UHFFFAOYSA-N
hydrogen pyrrolidine chloride
31268
71.12
Liquid
-63°C
86°C to 88°C
99%
C4H10ClN
MFCD00005249
102395
14,8015
Miscible with alcohol,ether,chloroform and water.
[H+].[Cl-].C1CCNC1
107.58
CHEBI:33135
99%
Pyrrolidine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H301-H314-H318-H332
Highly flammable liquid and vapour.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.

P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H302+H332-H314

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PYRROLIDINE

missing translation for 'einecsNumber' : 204-648-7

missing translation for 'rtecsNumber' : UX9650000

missing translation for 'tsca' : Yes

missing translation for 'storageNote' : Air sensitive

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.