missing translation for 'onlineSavingsMsg'
Learn More

Pyrrolidine, 99+%

Product Code. 10761414 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
100 mL
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
100mL
2.5 L
250mL
5mL
This item is not returnable. View return policy

Product Code. 10761414

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123-75-1 | C4H10ClN | 107.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-75-1
Molecular Formula C4H10ClN
Molecular Weight (g/mol) 107.58
MDL Number MFCD00005249
InChI Key FCLZCOCSZQNREK-UHFFFAOYSA-N
Synonym tetrahydropyrrole, tetrahydro pyrrole, azacyclopentane, azolidine, tetramethylenimine, butylenimine, perhydropyrrole, prolamine, 1-azacyclopentane, tetramethyleneimine
PubChem CID 31268
ChEBI CHEBI:33135
IUPAC Name hydrogen pyrrolidine chloride
SMILES [H+].[Cl-].C1CCNC1

Specifications

Melting Point -63.0°C
Density 0.8660g/mL
Boiling Point 86.0°C to 88.0°C
Flash Point 3°C
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4421 to 1.4441
Quantity 1 L
Beilstein 20,4
Fieser 01,972; 02,354; 03,240; 07,309; 08,428; 13,14
Merck Index 15,8129
Specific Gravity 0.866
Solubility Information Solubility in water: freely soluble. Other solubilities: soluble in alcohol, ether and chloroform, soluble in most organic solvents
Viscosity 0.94 mPa.s (20°C)
Formula Weight 71.11
Percent Purity 99+%
Chemical Name or Material Pyrrolidine, 99+%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.