missing translation for 'onlineSavingsMsg'
Learn More

Pyruvic acid, 98%, extra pure, Thermo Scientific Chemicals

Product Code. 10677292
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-17-3
Molecular Formula C3H4O3
Molecular Weight (g/mol) 88.06
MDL Number MFCD00002585
InChI Key LCTONWCANYUPML-UHFFFAOYSA-N
Synonym pyruvic acid, pyroracemic acid, 2-oxopropionic acid, acetylformic acid, alpha-ketopropionic acid, 2-ketopropionic acid, 2-oxopropanoate, propanoic acid, 2-oxo, 2-oxo-propionic acid, 2-oxopropansaeure
PubChem CID 1060
ChEBI CHEBI:32816
IUPAC Name 2-oxopropanoic acid
SMILES CC(=O)C(O)=O

Specifications

Melting Point 11.8°C
Color Amber or Colorless to Yellow
Density 1.2500g/mL
Boiling Point 165.0°C
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3COCO2H
Refractive Index 1.4260 to 1.4300
Quantity 100 g
Beilstein 03, 608
Fieser 01,974
Merck Index 15, 8135
Specific Gravity 1.25
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol and ether
Formula Weight 88.06
Percent Purity 98%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Pyruvic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Pyruvic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'