missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ pZerve Cryopreservation Solution

A cryopreservation solution that does not contain dimethyl sulfoxide (DMSO) and serum.

Brand:  MP Biomedicals™ 0920303M2

828.52 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.


 View more versions of this product

Product Code. 15757879

  • 891.00 € / 60mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

pZerve has been successfully used with a variety of cell types including mast cells, dendritic cells, and mesenchymal cells. It can be used for cells cultured in serum-free medium or with serum-containing medium.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at 2° to 8°C
Solution
Cryopreservation
pZerve Cryopreservation Media
Cryopreservation Media
60 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.