Learn More
Asecos™ Q-CLASSIC-90 Two-Door Light Gray Safety Storage Cabinets
For the safe and approved storage of flammable hazardous substances in working areas.
Brand:  Asecos™ 3000104133647
 View more versions of this product
Product Code. 15870448
  • 4250.00 € / Each
Explore more special offers
Description
Description
  • Extremely robust design with scratch-proof surface
  • Triple-hinged doors
  • Safety elements assembled outside the storage compartment for increased protection against corrosion
  • Integrated transport base for simple in-house transport
  • Cabinet can be transported upright through doors
  • Adjustable feet in base allow the safe and fast compensation of minor surface unevenness
  • Modern bicolor design - choice of seven different door colors
  • Language-free labelling for optimum international usability
  • Doors lockable with cylinder lock, cabinets can be integrated by the customer in an existing master key system
  • Locking status indicator (red/green) signals the locking status
  • Integrated air ducts ready for connection (DN 75) to a technical exhaust system
  • Visual inspection of the exhaust air function in the exhaust grille (rotating fan impeller)
  • Available with different combinations of drawers, height-adjustable shelves and bottom collecting sumps for leakages
  • In accordance with EN 14470-1 and TRGS 510
  • Fire resistance of 90 minutes
Specifications
Specifications
EN 14470-1
Triple-hinged
4 x 60 kg/29 L drawers
505 kg
Safety storage cabinet
615 x 1193 x 1953 mm
Storage of flammable hazardous substances
2
Light Gray