Learn More
QC Standard 21 For Environmental Analysis, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
358.00 € - 626.00 €
Specifications
Chemical Name or Material | ICP Quality Control Standard 21 |
---|---|
Packaging | Triple-leached LDPE bottle |
Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
Grade | Environmental |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11445938
|
SPEX Certiprep
QC-21 |
125 mL | Triple-leached LDPE bottle | N/A | |||||
11465938
|
SPEX Certiprep
QC-21-500 |
500 mL | Triple-leached LDPE bottle |
626.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
As | 100μg/mL |
Be | 100μg/mL |
Ca | 100μg/mL |
Cd | 100μg/mL |
Co | 100μg/mL |
Cr | 100μg/mL |
Cu | 100μg/mL |
Fe | 100μg/mL |
Li | 100μg/mL |
Mg | 100μg/mL |
Mn | 100μg/mL |
Mo | 100μg/mL |
Ni | 100μg/mL |
Pb | 100μg/mL |
Sb | 100μg/mL |
Se | 100μg/mL |
Sr | 100μg/mL |
Ti | 100μg/mL |
Tl | 100μg/mL |
V | 100μg/mL |
Zn | 100μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
ICP Quality Control Standard 21 | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Environmental |
Triple-leached LDPE bottle | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
5% HNO3/tr Tart. Acid/tr HF |
Safety and Handling
Causes severe skin burns and eye damage
missing translation for 'dotInformation' : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.