Learn More
Axygen™ Qiagen™ Robotic Tips
Compatible with Qiagen liquid handling systems and Qiagen BioRobot Series
Brand: Axygen™ QT-1100-CBK-R
This item is not returnable.
View return policy
Description
Available in volumes of 250, 300, 1000 and 1100 μL.
- Clear and conductive black tips available
- Choice of Presterilized, Nonsterilized, Filtered, Non-Filtered and Conductive tips
Specifications
1100 μL | |
Non-sterile | |
Non-ANSI | |
Non-pyrogenic, liquid level sensing, used for stringent specifications under strict process controls | |
Automation, Round | |
Polypropylene |
Non-filtered | |
Black | |
DNase, Rnase Free | |
Rack | |
Qiagen BioRobot |