Learn More
Axygen™ Qiagen™ Robotic Tips
GREENER_CHOICE
Compatible with Qiagen liquid handling systems and Qiagen BioRobot Series
Brand:  Axygen™ QTF-1100-CBK-R-S
 View more versions of this product
Product Code. 13417699
  • 376.00 € / Pack of 1536
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Available in volumes of 250, 300, 1000 and 1100 μL.
  • Clear and conductive black tips available
  • Choice of Presterilized, Nonsterilized, Filtered, Non-Filtered and Conductive tips
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1000 μL
Sterile
Non-ANSI
Non-pyrogenic, liquid level sensing, used for stringent specifications under strict process controls
Automation, Round
16 Racks/Cs.
Qiagen
Filtered
Black
DNase, Rnase Free
Rack
SBS Rack
Qiagen BioRobot
Polyethylene