Learn More
Thermo Scientific™ QMS™ Therapeutic Drug Monitoring (TDM) Assays
Utilize the precision, consistency and optimized performance of Thermo Scientific™ QMS™ TDM Assays, ready-to-use, liquid immunoassays.
Brand: Thermo Scientific™ 0373589
Description
Accurately measure therapeutic drug levels in serum and plasma with the Thermo Scientific™ QMS™ Therapeutic Drug Monitoring (TDM) Assays. These liquid, ready-to-use assays are a convenient alternative to more expensive and time-consuming technologies. QMS (Quantitative Microsphere System) immunoassays have excellent precision and accuracy, optimized performance on a wide range of general chemistry analyzers and lot-to-lot consistency.
Features:- Significantly reduces labor, time, and expense compared to HPLC or LCMS and other technologies
- Validated parameters assure lab-to-lab reproducibility
- Superb reagent stability
- Excellent correlation to reference methods
- Optimal sensitivity, precision and dynamic range
- Low cross-reactivity with metabolites and commonly co-administered drugs
- Low interference from endogenous substances
Includes:
Two reagent system, 22, 22mL

Specifications
2°C to 8°C | |
Vancomycin | |
Reagent |
Vancomycin Assay | |
Plasma, Serum | |
22 x 22 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.